Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   Lenz Laborglas
Multiple adapters
Supplier:  Lenz Laborglas
Certificates
Multiple adapters
Multiple adapters
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two sockets and one cone, standard ground joints and two parallel necks.
Documents
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,30 €
    29,30 €
    1 * 1 ST
       29,30 €
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,00 €
    35,00 €
    1 * 1 ST
       35,00 €
  • 24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,50 €
    41,50 €
    1 * 1 ST
       41,50 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,70 €
    60,70 €
    1 * 1 ST
       60,70 €