Adapters with screw thread and standard gound joint cone
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground joint cone.
DIN 12257
Frequently Bought Together