Orders LinkContactUsLinkComponent
Tanks, rectangular, with cover, Nalgene®
Tanks, rectangular, with cover, Nalgene®
   Sustainable Options Available
Tanks, rectangular, with cover, Nalgene®
   Sustainable Options Available
Certificates
Tanks, rectangular, with cover, Nalgene®
6 Options Available Below
Product Details & Documents
These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
  • Good stress-crack resistance
  • Suitable for use with many organic chemicals
  • Tanks can be modified with a factory-installed spigot
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-1921
  • 216-1922
  • 216-1923
  • 216-1924
  • 216-1925
  • NALG14200-0045
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 8 L
    200×200×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1921
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 20 L
    356×250×250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 593,00 €
    593,00 €
    1 * 1 ST
       593,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1922
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 30 L
    300×300×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 397,00 €
    397,00 €
    1 * 1 ST
       397,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1923
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 38 L
    457×300×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 496,00 €
    496,00 €
    1 * 1 ST
       496,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1924
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 L
    610×305×300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 723,00 €
    723,00 €
    1 * 1 ST
       723,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-1925
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 120 L
    610×457×450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 891,00 €
    891,00 €
    1 * 1 ST
       891,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG14200-0045
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact