Order Entry
Austria
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution, Supelco®
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution, Supelco®
Supplier:  Merck
Certificates
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.01 g/cm3 (20 °C)
  • pH:
    2.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together