Order Entry
Austria
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
Certificates
undefined
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution

Specification Test Results

  • Appearance:
    Dark red Liquid
  • Concentration:
    0.024 to 0.026 M (w/v in water)
  • Suitability:
    (for use as ind. for Fe) Passes test

Specification Test Results

Related Information

Frequently Bought Together