ContactUsLinkComponent
Venting filters for carboys, Nalgene®
Venting filters for carboys, Nalgene®
Catalog # 517-0001
   Low Manufacturing Impact
undefined
Venting filters for carboys, Nalgene®
Catalog # 517-0001
   Low Manufacturing Impact
Specifications
  • Filtration area:
    300 cm²
  • Pore size:
    0,2 µm
  • Seq:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    3
Specifications
About this item
These vent filters with hydrophobic PTFE membrane and PP housing are designed to maintain purified water stored in carboys. They can also be used for venting and solvent filtration applications.
  • Use up to five times in venting applications
  • 6 ml liquid hold-up volume