Nalgene® Bottles, Narrow Neck, Round, PPCO, Packaging, with Screw Caps
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, translucent with natural PP screw cap.
- Lightweight
- Very good chemical resistance
- Very good temperature resistance
- Linerless closure
Perform light-duty, general purpose applications where a heavier bottle is not required.
Economical and recyclable; designed for single use. Translucence allows for easy viewing of liquid levels.
Linerless caps ensure leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate the reagents.
Manufactured from non-toxic materials.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Specifications
- Catalog No:
- 215-2239
- 215-2240
- 215-2236
- 215-2237
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement