Tanks, cylindrical, with cover, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These cylindrical polypropylene tanks with cover can resist stress cracking and increase tolerance to many organic chemicals.
- An economic alternative to stainless-steel tanks
Ordering information: Factory-installed spigots and tank liners are sold separately.
Specifications
- Catalog No:
- 216-1906
- 216-1905
- 216-1903
- 216-1904
- 216-1902
- 216-1900
- 216-1901
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement