Order Entry
Austria
ContactUsLinkComponent
Thymolphthalein, Sigma-Aldrich®
Thymolphthalein, Sigma-Aldrich®
Catalog # SIAL114553-10G
Supplier:  Merck
CAS Number:  
Thymolphthalein, Sigma-Aldrich®
Catalog # SIAL114553-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :430.54 g/mol
Merck Index:12,09542
Melting Pt:253 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-729-7
CAS Number:125-20-2
Formula:C₂₈H₃₀O₄
MDL Number:MFCD00005909