Cation multi component standard 1 in water with a trace of nitric acid IC-MCA-01 for ion chromatography
Catalog # ACCUICMCA011
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:Cation multi component standard 1 in water with a trace of nitric acid IC-MCA-01 for ion chromatography