Order Entry
Austria
ContactUsLinkComponent
Methylene Blue zinc chloride double salt ~1 mol/mol H₂O for microscopy, Sigma-Aldrich®
Methylene Blue zinc chloride double salt ~1 mol/mol H₂O for microscopy, Sigma-Aldrich®
Catalog # SIAL66721-500G
Supplier:  Merck
CAS Number:  
Methylene Blue zinc chloride double salt ~1 mol/mol H₂O for microscopy, Sigma-Aldrich®
Catalog # SIAL66721-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • APPEARANCE (COLOR):
    Dark Brown to Very Dark Brown
  • APPEARANCE (FORM):
    Powder
  • TITRATION:
    80.0 - 120.0 %
  • TITRATION (METHOD):
    ARGENTOMETRIC AFTER OXYGEN COMBUSTION
  • THIN LAYER CHROMATOGR.:
    CORRESPONDS TO STANDARD
  • SOLUBILITY (COLOR):
    Dark Blue to Very Dark Blue
  • SOLUBILITY (METHOD):
    0.01G IN 10ML WATER
  • LOSS ON DRYING:
    ≤ 15 %
  • WAVELENGTH (1) (UV):
    656 - 666 nm
  • EXTINCTION (1%/1CM) (1) (UV):
    ≥ 1200
  • SOLVENT (UV):
    WATER

Specification Test Results

About this item

 :
 :388.00 g/mol (anhy)
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
EINECS:228-905-8
UN:2811
 :GHS06
CAS Number:26283-09-0
Formula:C₁₆H₁₈ClN₃S·0.5ZnCl₂·xH₂O
MDL Number:MFCD00150006