Order Entry
Austria
ContactUsLinkComponent
 
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98.0% (by titrimetric analysis) for complexometry, Fluka™
Catalog # 03779-50G
CAS Number:  
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98.0% (by titrimetric analysis) for complexometry, Fluka™
Catalog # 03779-50G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay (complexometric):
    97.0 - 101.0 %
  • Assay (alkalimetric):
    98.0 - 102.0 %
  • Loss on drying (105°C):
    Max. 2.0 %
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Sulfated ash:
    Max. 0.1 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 200 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 400 ppm

Specification Test Results

About this item

 :
Merck Index:13,03560
Melting Pt:241-250 °C
 :380.35 g/mol
Melting Pt:241…250 °C
Storage Temperature:Ambient
EINECS:200-651-2
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291