Order Entry
Austria
ContactUsLinkComponent
 
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 98%
 
undefined
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 98%
 

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    240 to 244 °C
  • Titration excess NaOH:
    ≥97.5 %
  • HPLC:
    ≥98.0 %

 

 

Frequently Bought Together