ContactUsLinkComponent
VWR® Capillary Tubes for Melting point Determinations
VWR® Capillary Tubes for Melting point Determinations
Catalog # 321242C
Supplier:  VWR Chemicals
undefined
VWR® Capillary Tubes for Melting point Determinations
Catalog # 321242C
Supplier:  VWR Chemicals
Specifications
  • Ext.Ø:
    ~1 mm
  • Description:
    Capillary tubes, hard glass
  • Length:
    100 mm
  • Environmentally Preferable:
  • Pk:
    100
Specifications
About this item
Hard glass.
  • Open both ends