Accessories for dissolved oxygen probe, StirrOx® G
: WTW
Frequently Bought Together
VWR® pHenomenal® pH Electrodes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Control seal, close-it tape
: BURKLE
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
Luminol (3-Aminophthalhydrazide) 98%, pure
: Thermo Fisher Scientific
BOD bottles, WHEATON®
: DWK Life Sciences
Caffeine ≥99.7%
: Thermo Fisher Scientific
Inlet liners, topaz
: Restek
Cellulose, microcrystalline ∼ Ø 90 µm, extra pure
: Thermo Fisher Scientific
Cut resistant gloves, ROCK CUT, TRIUS®
: TRIUS SAFETY