Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant FLT-3 (CTGrade GMP) (from E. coli)
New Product
 
Human Recombinant FLT-3 (CTGrade GMP) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.05
  • Biological activity:
    Bioactivity assay: Proliferation of OCI-Aml5, ED50 ≤5 ng/ml, bioactivity ≥1,6×10³ IU/µg
  • Gene ID:
    P49771
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Flt3L|Fms-related tyrosine kinase 3 ligand
  • Purity:
    ≥97%
  • Molecular weight:
    17,7 kDa
  • Sequence:
    MTQDCSFQHS PISSDFAVKI RELSDYLLQD YPVTVASNLQ DEELCGGLWR LVLAQRWMER LKTVAGSKMQ GLLERVNTEI HFVTKCAFQP PPSCLRFVQT NISRLLQETS EQLVALKPWI TRQNFSRCLE LQCQPDSSTL PPPWSPRPLE ATAPT
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

 

Frequently Bought Together