Order Entry
Austria
ContactUsLinkComponent
Human/Mouse Recombinant/M NT-3 (Animal free) (from E. coli)
New Product
Certificates
Human/Mouse Recombinant/M NT-3 (Animal free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
  • Storage conditions:
    At or below −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Proliferation of a neuroblastoma cell line stably expressing TrkB (BR6), ED50 ≤20 ng/ml, bioactivity ≥5,0×10⁴ units/mg
  • Gene ID:
    P20783/P20181
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    nerve growth factor 2 (NGF-2)|HGNF|neurotrophic factor|NT3|Neurotrophin 3
  • Purity:
    ≥95%
  • Molecular weight:
    13,8/27,5 kDa
  • Sequence:
    MYAEHKSHRG EYSVCDSESL WVTDKSSAID IRGHQVTVLG EIKTGNSPVK QYFYETRCKE ARPVKNGCRG IDDKHWNSQC KTSQTYVRAL TSENNKLVGW RWIRIDTSCV CALSRKIGRT
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together