Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant IL-4 (CTGrade GMP) (from E. coli)
New Product
 
Human Recombinant IL-4 (CTGrade GMP) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,05
  • Biological activity:
    Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥1,4x10⁴ IU/µg
  • Gene ID:
    P05112
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    BCGF|B-cell stimulatory factor-1|BCDF|BSF-1
  • Protein/peptide name:
    CTGrade GMP IL-4
  • Purity:
    ≥98%
  • Molecular weight:
    15,1 kDa
  • Sequence:
    MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS
  • Endotoxin level:
    Low
  • Formulation:
    20 mM Sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

 

Frequently Bought Together