Order Entry
Austria
ContactUsLinkComponent
Human Recombinant TPO (CHO) (from CHO cells)
New Product
Certificates
Human Recombinant TPO (CHO) (from CHO cells)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: MO7e cell proliferation, ED₅₀ ≤15 ng/ml, Bioactivity ≥6,7×10⁴ units/mg
  • Gene ID:
    P40225
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    c-MPL ligand|mgDF|Megakaryocyte Colony Stimulating Factor
  • Protein/peptide name:
    TPO (CHO)
  • Purity:
    ≥90%
  • Molecular weight:
    36,3 kDa
  • Sequence:
    SPAPPACDLR VLSKLLRDSH VLHSRLSQCP EVHPLPTPVL LPAVDFSLGE WKTQMEETKA QDILGAVTLL LEGVMAARGQ LGPTCLSSLL GQLSGQVRLL LGALQSLLGT QLPPQGRTTA HKDPNAIFLS FQHLLRGKVR FLMLVGGSTL CVRRAPPTTA VPSRTSLVLT LNELPNRTSG LLETNFTASA RTTGSGLLKW QQGFRAKIPG LLNQTSRSLD QIPGYLNRIH ELLNGTRGLF PGPSRRTLGA PDISSGTSDT GSLPPNLQPG YSPSPTHPPT GQYTLFPLPP TLPTPVVQLH PLLPDPSAPT PTPTSPLLNT SYTHSQNLSQ EGHHHHHH
  • Endotoxin level:
    Low
  • Formulation:
    5 mM Sodium phosphate, 2 mM L-Arginine, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together