ContactUsLinkComponent
Centrifugal Filter Units, with Microporous Hydrophilic PTFE Membrane
Centrifugal Filter Units, with Microporous Hydrophilic PTFE Membrane
Supplier:  Merck
Certificates
undefined
Centrifugal Filter Units, with Microporous Hydrophilic PTFE Membrane
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFUFC30LH25
  • MILFUFC30LG25
  • Filtration area:
  • 0,2 cm²
  • 0,2 cm²
  • Max. RCF:
  • 5000 ×g
  • 5000 ×g
  • Dead volume:
  • 5 µl
  • 5 µl
  • Ø×H:
  • 10,6×45 mm
  • 10,6×45 mm
  • Sample volume:
  • max. 400 µl
  • max. 400 µl
  • Temperature:
  • 50 °C
  • 50 °C