Order Entry
Austria
ContactUsLinkComponent
Mouse/Rat Recombinant FGF-acidic (from E. coli)
New Product
Certificates
Mouse/Rat Recombinant FGF-acidic (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
    Mouse
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P61148 / P61149
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    ECGF-beta|Beta-endothelial growth factor|HBGF-1|FGF1
  • Protein/peptide name:
    FGF-acidic/GF-1
  • Purity:
    ≥95%
  • Molecular weight:
    15,9 kDa
  • Sequence:
    MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 75 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together