Steriflip® Bottle Top Vacuum Filtration Systems, Durapore® (PVDF) membrane
Supplier: Merck
Certificates
About this item
The Steriflip® filter unit performs vacuum assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries. It also eliminates messy pouring during sample transfer. Filters come in different pore sizes that are ideal for cell isolation.
- Inlet fittings are double lead thread with vaccum port
- Attaches to any standard 50 ml centrifuge tube, eliminating unnecesary materials
Max. temperature: 45 °C
Frequently Bought Together