Ideal for sterilising tissue culture media, buffers, or other aqueous solutions. The no tip, tapered receiver flask and compact profile of the Stericup® design improve stability and make gripping the receiver easier.
- Connections: Hose barb vacuum inlet
- Max. operating temperature: 45 °C
- Sterile
The bottom of the flask is slightly recessed so that capped flasks can be stacked for storage. A small tab inside the funnel secures a prefilter in place during filtration. For added protection against contamination, the threads of the bottle cap are slightly recessed in case the cap accidentally touches a work surface.
Stericup® filter cups combine a filter unit with a receiver flask and cap for processing and storage.
Steritop™ bottle top filter units can be used with bottles having either a narrow mouth (33 mm) or a wide mouth (45 mm) opening.
- Catalog No:
- 515-0023
- MILFS2GPT10RE
- MILFS2VPU11RE
- MILFS2VPU02RE
- 515-0022
- 515-0032
- MILFS2GPT05RE
- MILFS2GPT01RE
- 515-0021
- 515-0031
- MILFS2GPT02RE
- Housing:
- Polystyrene
- Polystyrene
- Polystyrene
- Polystyrene