ContactUsLinkComponent
Millicell® EZ Slide Holder
Millicell® EZ Slide Holder
Catalog # MILFPEZXMSH01
Supplier:  Merck
undefined
Millicell® EZ Slide Holder
Catalog # MILFPEZXMSH01
Supplier:  Merck
Specifications
  • Description:
    Microscope slide holder for 4 and 8 well units, holds two slides, L×W×H: 128×86×147/17/95 mm, black
  • Material:
    PS
  • Seq:
    05
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Microscope slide holder for 4 and 8 well units, holds two slides.
  • Shipped in ambient temperature
Techniques: cell based assay: suitable cell culture; mammalian: suitable immunocytochemistry: suitable