Order Entry
Austria
ContactUsLinkComponent
Solvent Filtering Dispenser
Solvent Filtering Dispenser
Catalog # MILFXX6602500
Supplier:  Merck
undefined
Solvent Filtering Dispenser
Catalog # MILFXX6602500
Supplier:  Merck

Specifications

  • Volume:
    1 L
  • Description:
    Solvent filtering dispenser, 39 mm² filtration area, Ø 25 mm, 318 mm height
  • Pk:
    1

Specifications

About this item

Ultraclean and dispense small volumes of solvent by squeeze-bottle action. This hand-pressure operated dispenser features a filter holder fitted with a delivery tube. Use to direct filtered solvent against surfaces.

  • Integrated hand pump eliminates the need for external pump
  • Holds up to 1 L of solvent

Applications: Solvent filtration prior to contamination analysis, solvent rinsing of machined parts and collection containers.

Components: TFE gasket, PTFE seal, PTFE tubing, borosilicate glass flask, glass elbow(s), glass nozzle (for dispensing), neoprene bulb, neoprene stopper, stainless steel holder (for filter).