Order Entry
Austria
ContactUsLinkComponent
Luminata™ Western HRP Substrates Samples for Antibodies of All Affinities
Luminata™ Western HRP Substrates Samples for Antibodies of All Affinities
Catalog # MILFWBLUM0100
Supplier:  Merck
CAS Number:  
undefined
Luminata™ Western HRP Substrates Samples for Antibodies of All Affinities
Catalog # MILFWBLUM0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Samples of all three Luminata western HRP substrates for antibodies of all affinities

Specifications

About this item

Western blotting, binding assays, amino acid analysis, N-terminal protein sequencing, dot/slot blotting, glycoprotein visualisation, lipopolysaccharide analysis.

  • Detection method: Chemiluminescence
Delivery: Components includes:
Luminata classico western HRP substrate, 100 ml (MILFWBLUC0100)
Luminata crescendo western HRP substrate, 100 ml (MILFWBLUR0100)
Luminata forte western HRP substrate, 100 ml (MILFWBLUF0100)