Order Entry
Austria
ContactUsLinkComponent
MultiScreen® HTS Filter Plate, Plasmid
MultiScreen® HTS Filter Plate, Plasmid
Catalog # MILFMSNUPSD50
Supplier:  Merck
MultiScreen® HTS Filter Plate, Plasmid
Catalog # MILFMSNUPSD50
Supplier:  Merck

Specifications

  • No. of wells:
    96
  • Pore size:
    Proprietary
  • Sterile:
    -
  • Colour:
    Clear
  • Material:
    Polystyrene
  • Max. working volume/well:
    50 - 250 μl sample volume (per well)
  • Pk:
    50

Specifications

About this item

Multiscreen HTS filter plate, plasmid also called as Montage Plasmid Miniprep.

  • Automation compatible
  • Suitable for DNA purification
  • Shipped in: Ambient
  • Quality level: 400

Application: Research category and nucleic acid