Millipore® Steripak Pressure Filters
Supplier: Merck
Certificates
About this item
Steripak-GP filter units are designed for larger scale pressure-driven filtration of tissue culture media with or without serum. They are single use and are not autoclavable. The filter units come in two sizes, with the smaller unit able to filter up to 10 L and the larger up to 20 L.
- Millipore Express PLUS membrane for fastest flow rates and low protein binding
- Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
- 10 and 20 L capacity sizes for maximum laboratory-scale filtration options
- Manually operated top air vent to prevent air lock stop of flow
- Filling bell for cleaner filling environment around the opening of the receptacle bottle
Both sizes utilize the low protein binding, fast flow 0,22 µm Millipore Express membrane. Each unit has an integral filling bell for bottle-top use and a manually operated upstream air vent to prevent membrane air locking. They are supplied sterile and ready to connect to a pump or pressure vessel (not included).
Caution: For research use only.
Specifications
- Catalog No:
- MILFSPGPM10RJ
- MILFSPGPM20RJ
- Filtration area:
- 100 cm²
- 200 cm²
- Filter type:
- Pressure
- Pressure