ContactUsLinkComponent
SNAP i.d. 2.0 Protein Detection Systems Mini
SNAP i.d. 2.0 Protein Detection Systems Mini
Catalog # MILFTISNAP2MINI
Supplier:  Merck
undefined
SNAP i.d. 2.0 Protein Detection Systems Mini
Catalog # MILFTISNAP2MINI
Supplier:  Merck
Specifications
  • Detector:
    Chemiluminescence fluorescence chromogenic
  • Description:
    SNAP i.d. 2.0 Protein detection system
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Ordering information: This system includes the SNAP i.d. 2.0™ base, two mini blot holding frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.