Order Entry
Austria
Orders LinkContactUsLinkComponent
Multiscreen® SEQ₃₈₄ Filter Plates

Multiscreen® SEQ₃₈₄ Filter Plates

Supplier:   Merck
Multiscreen® SEQ₃₈₄ Filter Plates
Supplier:  Merck
Certificates
Multiscreen® SEQ₃₈₄ Filter Plates

2 Options Available Below

Product Details & Documents

The MultiScreen384-SEQ filter plate is suited for use with most liquid handling systems. The filter plate operates by vacuum filtration enabling top-loading and recovery of sample. Our plate is designed with the needs of the high throughput laboratory in mind, meets SBS standards and is ideally suited for 'walk away' automation.

  • Automation friendly: Design and protocol saves time, money and improves reproducibility
  • Efficient dye terminator & salt removal: Recovers highly purified sequencing products for long, accurate sequencing reads
  • Eliminates dry-down step: Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
  • Saves reagents: plates suitable for use with ¹/₁₆ sequencing reactions
  • Proprietary membrane
  • Color code: Clear
  • Automation compatible: No

Application: Research category, assays, general filtration and nucleic acid.

Packaging: Does not include reagents.

Product Family Options

Product Information

  • Colour
    Recommended working volume
    No. of wells
    Sterile
    Availability
    Price
  • MILFS384SEQ10
    Multiscreen® SEQ₃₈₄ filter plate
    Multiscreen® SEQ₃₈₄ filter plate
    Clear
    100 µl
    384
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • MILFS384SEQ50
    Multiscreen® SEQ₃₈₄ filter plate
    Multiscreen® SEQ₃₈₄ filter plate
    Clear
    100 µl
    384
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST