Order Entry
Austria
ContactUsLinkComponent
Millex® PVDF Syringe Filter
Millex® PVDF Syringe Filter
Supplier:  Merck
Certificates
undefined
Millex® PVDF Syringe Filter
Supplier:  Merck

Specifications

  • Catalog No:
  • MILFSLGVR33RB
  • MILFSLVVR33RS
  • MILFSLSV025NB
  • MILFSLGVR33RS
  • 514-0437
  • MILFSLHVR33RS
  • 514-0425
  • MILFSLHVR33RB
  • MILFSLHVR13SL
  • Ø:
  • 33 mm
  • 33 mm
  • 25 mm
  • 33 mm
  • 4 mm
  • 33 mm
  • 4 mm
  • 33 mm
  • 13 mm
  • Hold-up volume:
  • <100 μl
  • < 10 μl hydrophilic
  • < 10 μl hydrophilic
  • Max. operating pressure:
  • 10,34 bar
  • 10,34 bar
  • 5,2 bar max. inlet pressure (75 psi)
  • 10,34 bar
  • 14 bar
  • 10,34 bar
  • 14 bar
  • 10,34 bar
  • 7 bar
  • Max. temperature:
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • Water flow rate:
  • ≥40 ml/min
  • ≥25 ml/min
  • ≥40 ml/min
  • ≥300 ml/min
  • ≥300 ml/min

Specifications

Related Information

Frequently Bought Together