Order Entry
Austria
ContactUsLinkComponent
Amicon® Pro Purification System
Amicon® Pro Purification System
Catalog # MILFACS500024
Supplier:  Merck
CAS Number:  
undefined
Amicon® Pro Purification System
Catalog # MILFACS500024
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Amicon® Pro Purification system
  • Volume:
    10 ml

Specifications

About this item

The Amicon® Pro purification system - A centrifugal tool collapsing protein purifcation with concentration and buffer exchange.

  • Minimum final concentrate volume: 15 µl
  • Non sterile
  • Cap: Polyethylene
  • Frit: Polypropylene/polyethylene
  • Colour code: Clear with pink cap

The Amicon® Pro purification system 24 pack includes 24 exchange devices and 24 collection tubes with cap.

Applications include: Protein purification, protein concentration, buffer exchange, protein enrichment, antibody labeling.

Ø×Length: 29,7×120 mm