About this item
The Montage® SEQ96 sequencing reaction cleanup kit provides the filtration plate and solution necessary to remove contaminating salts and unincorporated dye terminators from DNA sequencing reactions.
- Colour code: Blue
- Technique: DNA sequencing
- Plate size: 96 wells
- Application: Genomic analysis
- Non sterile
The vacuum-based size-exclusion separation platform eliminates the need for gel filtration or bind-wash elute columns that require centrifugation. The entire protocol requires very little time to process typically less than 10 minutes. In addition, the optimised filtration area enables reaction miniaturisation and reagent reduction, resulting in lower DNA sequencing costs. Montage® SEQ96 sequencing reaction clean-up kit consists of 96 well SEQ plate and an injection solution.
Application: Purification of polymerase chain reaction (PCR) products and to remove unincorporated primers and dye terminators for mutational analysis of Krüppel-like factor 4 (KLF4), purification of sequencing reactions, 16S rDNA sequence analysis to remove unincorporated dye terminators from the sequencing reaction.