ContactUsLinkComponent
Millicup™-FLEX Disposable Filtration Kits
Millicup™-FLEX Disposable Filtration Kits
Supplier:  Merck
Certificates
undefined
Millicup™-FLEX Disposable Filtration Kits
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFMCFLX4710
  • MILFMCFLX4702
  • Type:
  • Bottle top filter
  • Bottle top filter
  • Sample volume:
  • 250 ml
  • 250 ml
  • Autoclavable:
  • Not autoclavable
  • Not autoclavable
  • Membrane:
  • Cut disc filters, Ø 47 mm
  • Cut disc filters, Ø 47 mm