Amicon™ XM Ultrafiltration Membranes Dics Filters
Artikelnr. MILF14112AM
Supplier: Merck
Specifications
- Ø:25 mm
- MWCO:50,000 NMWL
- Type:Ultrafiltration filter disc
- Description:Ultrafiltration membrane, disc, Amicon™ XM-50
- Seq:01
- Pk:10
Specifications
About this item
Ultrafiltration membranes - Amicon™ XM polyacrylonitrile/PVC co-polymer filters. High-recovery membranes for stirred cells.
- Regenerated cellulose membranes
- For concentration of hydrophobic samples or to desalt extremely dilute solutions
Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules.