Millex Samplicity® Filters
: Merck
Millex Samplicity® Filters for the Samplicity® filtration system.
- Samplicity® Filtration System enables vacuum driven filtration of up to eight samples at once, directly into HPLC vials
- Filtration for HPLC sample prep without syringes
- Millex Samplicity® Filters provide integrated funnels and filters for easiest sample loading
- Available in 0,20 µm and 0,45 µm pore sizes with hydrophilic PTFE and Durapore® PVDF membranes
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC.
Frequently Bought Together