Order Entry
Austria
ContactUsLinkComponent
30 Minute IHC DAB Detection System
30 Minute IHC DAB Detection System
Catalog # ANTIA320597-100
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
30 Minute IHC DAB Detection System
Catalog # ANTIA320597-100
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    DAB detection system

Specifications

About this item

This 30 Minute IHC DAB Detection System is a powerful intensification system that significantly enhances chromogenic signals. This system can be integrated into standard immunohistochemistry (IHC) staining methods and results in a significant increase in sensitivity compared to standard IHC detection methods, while maintaining similar specificity.

  • Fast results: Delivers results in just 30 minutes, enhancing laboratory efficiency
  • Enzyme-mediated reaction: Utilises horseradish peroxidase (HRP) to catalyse key reactions
  • Dual linkers for precision:
  • Linker 1: Introduces HRP into the system
  • Linker 2: Introduces Biotin for targeted binding
  • Enhanced detection with tracer: Tracer introduces both Streptavidin and additional HRP for amplified signal detection
  • Versatile detection: linkers can be detected using Streptavidin Peroxidase conjugate followed by a DAB chromogen solution
  • High sensitivity: HRP catalyses the reaction, allowing for clear visualisation of the target on tissue sections or cell preparations

All reagent components are formulated without Sodium azide or Thimerosol as preservatives. The reagents are provided in ready-to-use, with the exception of Stable DAB/Plus.

Caution: This product is for research use only. It is not intended for diagnostic or therapeutic use.