Order Entry
Austria
ContactUsLinkComponent
AAV1/2-CMV/CBA-Null/Empty AAV Virus Vector
AAV1/2-CMV/CBA-Null/Empty AAV Virus Vector
Catalog # GD1004-RV
New Product
CAS Number:  
AAV1/2-CMV/CBA-Null/Empty AAV Virus Vector
Catalog # GD1004-RV
New Product
CAS Number:  

Specifications

  • Concentration:
    5×10¹² vg/ml
  • Pk:
    25 µl
  • Vector details:
    AAV1/2-CMV-CBA-Null/Empty-WPRE-BGH-polyA

Specifications

About this item

AAV1/2 Vector with CMV promoter, Null/Empty, WPRE, bovine growth hormone (BGH), and polyadenylation sequence.

  • CMV Promoter
  • AAV1/2 Serotype
  • PBS with 1 mM MgCl₂ and 0.001% Lutrol F68 storage buffer

The CMV/CBA promoter consists of the chicken beta-actin promoter hybridised with the CMV immediate early enhancer sequence and is highly efficient in most tissue types. The Woodchuck post-transcriptional
regulatory element (WPRE) and the presence of a bovine growth hormone (BGH) polyadenylation sequence ensures high transcription following transduction.

Shipped in dry ice.