Order Entry
Austria
ContactUsLinkComponent
 
AAV1/2-CMV/CBA-Human-A53T-alpha-SYN AAV Vector
Catalog # GD1001-RV
New Product
CAS Number:  
AAV1/2-CMV/CBA-Human-A53T-alpha-SYN AAV Vector
Catalog # GD1001-RV
New Product
CAS Number:  

Specifications

  • Concentration:
    5,1×10¹² vg/ml
  • Pk:
    25 µl
  • Storage conditions:
    –80 °C
  • Vector details:
    AAV1/2-CMV/CBA-Human-A53T-alpha-synuclein-WPRE-BGH-polyA

Specifications

About this item

AAV1/2 Vector with CMV promoter, human A53T alpha-synuclein, WPRE, bovine growth hormone (BGH), and polyadenylation sequence.

  • CMV promoter
  • AAV1/2 Serotype
  • PBS with 1 mM MgCl₂ and 0,001% Lutrol F68 storage buffer

The CMV/CBA promoter consists of the chicken beta-actin promoter hybridised with the CMV immediate early enhancer sequence and is highly efficient in most tissue types. The Woodchuck post-transcriptional regulatory element (WPRE) and the presence of a bovine growth hormone (BGH) polyadenylation sequence ensures high transcription following transduction.

Shipped in dry ice.