Showa XC500 Cut Resistant Gloves
Supplier: Showa
New Product
Certificates
About this item
The new ultra-light XC500 combines high cut-resistance level E/A5 provided by a seamless HPPE/Stainless steel/Polyester liner with a durable, abrasive-resistant polyurethane palm coating usable in various industrial applications.
- Ergonomic, flexible, lightweight and breathable
- Increased dexterity
- Seamless knit and form-fitting
- Abrasion-resistant with rough grip
- HPPE and stainless steel reinforced
- Polyurethane palm coating
- 12 Pair per polybag, 6 polybags per case, 72 pair per case
Trades and applications: Light assembly of oil-coated pieces, Maintenance, Mechanical and engineering, Repair glass and windows and Component assembly.
Certifications: EN 388:2016+A1:2018, ANSI/ISEA 105-2016 CUT.
Frequently Bought Together