Buffered NaCl Peptone Solution pH 7,0
New Product
Buffered NaCl Peptone Solution pH 7,0 is used for dissolving, suspending and diluting test samples. Its composition conforms to recommendations in the Harmonised Pharmacopoeias for the microbiological examination of non-sterile products.
: This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics.
: For professional use only.
- Storage temperature:10...25 °C
Frequently Bought Together