SAVIEW® PLUS HRP Reagent
Artikelnr. ENZOENZACC1020150
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:150 Tests
- Description:SAVIEW® PLUS HRP reagent
Specifications
About this item
High sensitivity, low background streptavidin-based nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
- Ready-to-use reagent
- Streptavidin-based detection with reduced carbohydrate and biotin background
- Suitable for both manual and automated platforms
- Suitable for custom multiplex IHC protocols
- The Streptavidin blocker/diluent further reduces background (sold separately)
- Customise with HIGHDEF® chromogens of your choice (sold separately)
SAVIEW® PLUS HRP reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW® PLUS HRP reagent can be used for in situ hybridisation detection of biotinylated DNA/RNA probes.
Delivery: 150 tests × 150 µl (22,5 ml total volume).