Order Entry
Austria
ContactUsLinkComponent
Screw Cap for Container
Screw Cap for Container
Catalog # SEMA10729
Supplier:  SEMADENI
New Product
Screw Cap for Container
Catalog # SEMA10729
Supplier:  SEMADENI
New Product

Specifications

  • Ø:
    29 mm
  • Colour:
    Transparent
  • Material:
    HDPE
  • Compatible with:
    Screw cap, Height: 16 mm
  • Pk:
    200

Specifications

About this item