Order Entry
Austria
Orders LinkContactUsLinkComponent
Newbox, PP

Newbox, PP

58,5 L, Grey, 600×400×320 mm, 2,8 kg, RAKO stacking containers
Catalog # SEMA12672
Supplier:   SEMADENI
Newbox, PP
Catalog # SEMA12672
Supplier:  SEMADENI
Newbox, PP

58,5 L, Grey, 600×400×320 mm, 2,8 kg, RAKO stacking containers

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The closed Euro-standard container of the Newbox range boasts a slim design. Thanks to its robustness and dimensional stability, the Newbox is a durable and economic product.

  • Temperature resistant up to 80 °C (short term up to 100 °C)
  • 100% compatible with RAKO stacking containers
  • With two and four handle openings

The containers are stackable and dimensionally matched with Euro pallets. Compared with the Logiline range, the Newbox range impresses with a somewhat slimmer and lighter design that is nevertheless robust and dimensionally stable.

Applications: With manual handling closed Newbox containers are used universally as transport containers or for the storage of the widest possible range of goods.

Specifications
  • Capacity:
    58,5 L
  • Colour:
    Grey
  • W×D×H:
    600×400×320 mm
  • Compatible with:
    RAKO stacking containers
  • Weight:
    2,8 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.