ContactUsLinkComponent
CryoGen® Internal Cap for Tubes, 2D
CryoGen® Internal Cap for Tubes, 2D
Catalog # BSGMCL2ARBIPS2D
Supplier:  BIOSIGMA
New Product
CryoGen® Internal Cap for Tubes, 2D
Catalog # BSGMCL2ARBIPS2D
Supplier:  BIOSIGMA
New Product
Specifications
  • Description:
    Internal cap with self standing bottom style, Height: 47,70 mm (±0,34 mm)
  • Size:
    2 ml
  • Pk:
    500
Specifications
About this item
Human DNA, DNase, RNase, Endotoxins, ATP, PCR inhibitors free certified.