Order Entry
Austria
ContactUsLinkComponent
Certipur® ICP Multi Element Standard Solution IV
Certipur® ICP Multi Element Standard Solution IV
Catalog # 1044980100.
Supplier:  Merck
New Product
CAS Number:  
Certipur® ICP Multi Element Standard Solution IV
Catalog # 1044980100.
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Matrix:
    Nitric acid dil.
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    ICP multi-element standard solution IV 23 elements
  • Hazard class description:
    Corrosive
  • UN No.:
    2031
  • GHS signal:
    Danger
  • Storage temperature:
    15...25 °C

Specifications

About this item

Certifications: This certified reference material (CRM) is produced in accordance with ISO 17034 and characterised in accordance with ISO/IEC 17025.