Order Entry
Austria
ContactUsLinkComponent
Cholesterol (synthetic), Synthechol®
Cholesterol (synthetic), Synthechol®
Catalog # 1.04672.0010
New Product
CAS Number:  
Cholesterol (synthetic), Synthechol®
Catalog # 1.04672.0010
New Product
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :386.66 g/mol
Boiling Pt:360 °C
Melting Pt:147...149 °C (lit.)
Density:1.067 g/ml at 25 °C (lit.)
 :M-Clarity™ quality level = MQ500 (more info.)
CAS Number:57-88-5
Formula:C₂₇H₄₆O
MDL Number:MFCD00003646