Order Entry
Austria
ContactUsLinkComponent
Acetobacter Agar Dehydrated Culture Media
Acetobacter Agar Dehydrated Culture Media
Catalog # HIMEM238-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Acetobacter Agar Dehydrated Culture Media
Catalog # HIMEM238-500G
Supplier:  HIMEDIA
New Product
CAS Number:  

Specifications

  • Pk:
    500 g
  • Description:
    Acetobacter agar (Glucose)

Specifications

About this item

Dehydrated culture media for enrichment, isolation, cultivation, and maintenance of microorganisms.

  • Microbiological culture media
  • Packaged in 500 g poly bottles

Application: Maintenance of glucose-positive Acetobacter species