Order Entry
Austria
ContactUsLinkComponent
Savant™ SpeedVac™ SPD130DLX Medium Capacity Concentrator for Combinatorial Chemistry Applications
Savant™ SpeedVac™ SPD130DLX Medium Capacity Concentrator for Combinatorial Chemistry Applications
Catalog # THSASPD130DLX-230
Supplier:  THERMO SAVANT
undefined
Savant™ SpeedVac™ SPD130DLX Medium Capacity Concentrator for Combinatorial Chemistry Applications
Catalog # THSASPD130DLX-230
Supplier:  THERMO SAVANT

Specifications

  • Description:
    SpeedVac™ SPD130DLX medium capacity modular vacuum concentrator, ambient, 35...80 °C in 5 °C increments, 230 V, 50 Hz
  • W×D×H:
    33×46×36 cm
  • Weight:
    26 kg
  • Pk:
    1

Specifications

About this item

The Thermo Scientific™ Savant™ SpeedVac™ SPD130DLX vacuum concentrator is a medium-capacity, vacuum concentrator with a modular design. It is compatible with a wide variety of solvents and offers resistance to TFA, DMSO, and other aggressive solvents used in combinatorial chemistry applications.

  • Proven reliability: Over 50 years of experience with the Savant SpeedVac family
  • Consistent drying time: Heated glass lid reduces drying times and prevents condensation
  • User-friendly: Intuitive interface with three preset and three user-defined programs for efficient operation
  • Safety assurance: Soft-close lid locks securely; hands-free operation with lid stay mechanism and audible alarm
  • Data transfer: USB port for downloading live run data enhances transferability

Our advanced solvent solutions are meticulously formulated to cater to a wide range of laboratory and industrial applications, including the removal of ethyl acetate from samples for CAT assays, the extraction of drug metabolites from solid-phase extracts, and the elution of analytes for evapouration processes. Featuring high-purity options such as Acetone, Chloroform, Ethyl Acetate, Hexane, and Methylene Chloride, our solvents ensure optimal performance, minimal interference, and maximum recovery of target compounds. These solvents are designed to deliver reliable, efficient, and reproducible results, making them indispensable in pharmaceutical, analytical, and various other scientific endeavors.