Savant™ SpeedVac™ SPD2030P1 Integrated Vacuum Concentrator System Kit
531-1495
- Description:SpeedVac™ SPD2030 integrated vacuum concentrator system kit, 35...80 °C in 5 °C increments, 230 V, 50 Hz
- Environmentally Preferable:
- W×D×H:79×71×42 cm
- Weight:98 kg
- Pk:1 KIT
Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 integrated SpeedVac™ systems and kits.
- Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
- Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
- Ease of use: Intuitive design with a bright display, three preset and three user-defined programs, and an oil-free vacuum source. USB port for data transfer
- Safety assurance: Soft-close glass lids lock securely for the safety of samples and technicians
Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 system is designed for medium capacity (i.e. 120×1,5 ml tubes, other formats). SpeedVac SPD2030 system allows higher capacity (i.e. 200×1,5 ml tubes, other formats).
Solvent and solvent combinations: Acetonitrile, ethanol, methanol, methylene chloride, water, other organic solvents
: SPD2030P1 Kit includes: Integrated SpeedVac system, 200×1,5 ml rotor (RH200-12), CryoCool™ 1 L heat transfer fluid, glass condensation flask (GCF400).